missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXPRESS One-Step Superscript™ qRT-PCR Kit, universal

Kit incorporates proven Invitrogen™ enzymes specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for probe-based qPCR experiments.

$2,045.00 - $8,590.00

Specifications

Detection Method Primer-probe
Format Tube
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
11781200
Encompass
View Documents Promotion Details
Invitrogen™
11781200
500 Reactions
Each for $2,045.00
Only null left
 
1178101K
Encompass
View Documents Promotion Details
Invitrogen™
1178101K
2500 Reactions
Each for $8,590.00
Only null left
 
Description

Description

  • Kit incorporates proven Invitrogen™ enzymes specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for probe-based qPCR experiments
  • This fast, one-step mix is designed to work with a variety of instruments’ default protocols, including Applied Biosystems and non-Applied Biosystem (see universal version) qPCR platforms
  • Fast-activating, antibody-mediated Platinum Taq DNA Polymerase for complete, rapid activation; provides high sensitivity and reproducibility
  • The One-Step SuperScript III Reverse Transcriptase module is a unique formulation to ensure high yield and less PCR inhibition
  • Flexible formats with separate or pre-mixed ROX are designed to work with default instrument protocols on a wide range of high-throughput fast instruments
  • Contains heat-labile UDG which significantly reduces carryover contamination risk with excellent activity from room temperature to approximately 40°C
  • Due to the thermostability of SuperScript III, the cDNA synthesis step is run at 50°C, at which temperature the heat-labile UDG is totally and irreversibly inactivated
  • Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at 4°C to 8°C
  • Store at -20°C for long-term storage
Specifications

Specifications

Primer-probe
1-step RT-qPCR
Fast or Standard
SuperScript
RNA
Tube
Taq DNA Polymerase
7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
qRT-PCR Kit
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.