missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ ExpiSf™ Protein Production Kit
Description
The ExpiSf Protein Production Kit is designed to significantly boost protein expression from high-density suspension cultures of Sf9 insect cells. The kit includes the chemically defined, animal origin-free ExpiSf Enhancer and ExpiSf CD Medium, which together power the highest possible level of protein expression from baculovirus-infected ExpiSf9 Cells.
The ExpiSf Protein Production Kit:
- Delivers greater than 80% more protein yield in high-density suspension Sf9 cell culture than ExpiSf CD Medium alone
- Includes ExpiSf Enhancer, which is specifically designed to work in concert with ExpiSf CD Medium to significantly boost protein expression from baculovirus-infected insect cell cultures
- Demonstrates direct scalability of baculovirus infection from culture volumes of 4 mL in deep-well plates to >3 L in shake flasks
The ExpiSf Protein Production Kit is specifically designed to work with ExpiSf9 Cells in the ExpiSf Expression System. The kit supplies sufficient ExpiSf Enhancer and ExpiSf CD Medium to infect 1 L, 10 L, or 50 L of culture. ExpiSf CD Medium is also available for purchase separately.
Specifications
Specifications
| Type | Media and reagents, ExpiSf CD Meidum and ExpiSf Enhancer |
| Quantity | 5 x 10 L (ExpiSf CD Medium), 5 x 40 mL (ExpiSf Enhancer) |
| Content And Storage | 5 x 10 L bag ExpiSf CD Medium, store in the dark at 2°C to 8°C; 5 x 40 mL ExpiSf Enhancer, store (at 2°C to 8°C; Shelf Life: 12 months from date of manufacture) |
| Classification | Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free |
| Compatible Cells | Insect Cells |
| Format | Liquid |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Expression Mechanism | Cell-Based Expression |
| Expression System | Baculovirus |
| Product Line | ExpiSf |