missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiCHO™ Expression System Kit

Catalog No. A29133
Encompass
Change view
Click to view available options
Cell Line:
CHO-S, SFM Adapted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Line
A29133 CHO-S, SFM Adapted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29133 Supplier Gibco™ Supplier No. A29133
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

2 vials frozen ExpiCHO-S™ cells, 1 bottle 1000mL ExpiCHO™ Expression Medium, 1 ExpiFectamine™ CHO transfection kit for 1 L of culture, 1 vial Antibody Expressing Positive Control Vector, 1 bottle 100mL OptiPRO™ SFM; store all components at +2° - +8° C, except frozen cells stored in liquid nitrogen

The ExpiCHO Expression System Kit combines the power of rapid, ultra-high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells.

The ExpiCHO Expression System Kit combines the power of rapid, ultra–high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells. It is based on high-density culture of ExpiCHO-S cells in ExpiCHO Expression Medium. Transient expression is powered by ExpiFectamine CHO transfection reagent, designed specifically for high-density CHO suspension culture, in combination with a specialized transfection enhancer and culture feed.

All components work together synergistically with expression yields of 1–3 g per L of transfected culture demonstrated for some antibody and non-antibody proteins.

The ExpiCHO Expression System enables:

  • Milligram to multigram yields of recombinant protein per liter of culture in 5–14 days post-transfection
  • Robust and reproducible transfection results, for greater confidence in results and ability to use CHO cells from research to discovery
  • Transfection efficiency of > 90% for high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
  • Use of one-third to one-half less plasmid DNA than other transfection reagents 
  • Flexible, reliable culture and transfection protocols with multiple options that readily fit into current transient expression workflows
  • Scalable expression for culture formats from  <1 mL in deep well plates up to 5 L when using Nalgene angled-bottom shake flasks

The components included in the ExpiCHO Expression System Kit are: 2 vials of frozen ExpiCHO-S cells, 1 L of ExpiCHO Expression Medium, 1 ExpiFectamine CHO Transfection Kit sufficient to transfect 1 L of culture, OptiPRO SFM, and an antibody-expressing positive control vector.

ExpiCHO-S cells, ExpiCHO Expression Medium, ExpiFectamine CHO Transfection Kit, and OptiPRO SFM may also be purchased separately.

All components of the system are animal origin–free.

Specifications

Type Expression System
Protein Tag Untagged
Delivery Type Transfection
Quantity 1 kit
Content And Storage • 2 vials frozen ExpiCHO-S cells
• 1000 mL ExpiCHO Expression Medium
• 1 ExpiFectamine CHO Transfection Kit for 1 L of culture
• 1 vial Antibody Expressing Positive Control Vector
• 100 mL OptiPRO SFM

Store all components at 2-8°C, except frozen cells which should be stored in liquid nitrogen.
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Cell Line CHO-S, SFM Adapted
Expression System Mammalian
Product Line ExpiCHO
For Use With (Application) Protein Production, Constitutive Expression
Promoter CMV
Show More Show Less
How can I learn more about commercial licensing of Freedom kits, ExpiCHO-S Cells (cGMP banked), CHO-S Cells (cGMP banked), and CHO DG44 Cells (cGMP banked)?

To inquire about a Commercial Production or Service License, please email: outlicensing@thermofisher.com.

When subculturing ExpiCHO the day before a transfection to 3-4 x 10E6/mL, would you recommend pelleting the cells and resuspending them in completely fresh media, or simply replacing some of the media to split them down to the desired cell density?

Before transfecting ExpiCHO cells, there is no need to change out the media during the subculturing. This would become expensive and the conditioned media is not necessary.

For how many passages would you recommend using ExpiCHO cells before replacing them with new cells that were frozen at earlier passages?

For optimal performance with respect to protein production, we recommend thawing ExpiCHO cells and letting them grow for 2-3 passages prior to transfection. Overall, we do not recommend using the cells beyond ˜20 passages.

With the ExpiCHO Expression System, can I shake faster or slower than recommended?

We have provided optimal shake speeds for various formats. For most flasks, there is a drop-off in expression when you go too fast or slow because of cell shear stress or insufficient aeration. The effect is sharper in plates where there is a sharper transition between static and moving fluid in wells.

How many days should I wait before harvesting my protein with the ExpiCHO system?

Optimal time to harvest protein will depend on the specific properties of the protein being expressed and the protocol chosen. Typical harvest times to reach maximum titers for the various protocols are as follows:
- Standard Protocol: 8-10 days post-transfection
- High Titer Protocol: 10-12 days post-transfection
- Max Titer Protocol: 12-14 days post-transfection

I didn't add the enhancer with the ExpiCHO system. Will I still get good protein expression?

The Enhancer is designed to work with the ExpiCHO system for maximal expression. If the enhancer is not added, there will be a greater than 50% loss in expression.

Can I use a different transfection reagent with the Expi-CHO Expression System?

The ExpiFectamine CHO Transfection Reagent, ExpiFectamine CHO Enhancer and ExpiCHO Feed are optimized to work together to provide maximal protein expression levels and are provided as a single kit for convenience. The ExpiFectamine CHO Transfection Reagent provides high transfection efficiency of high density cultures, superior to any other transfection reagent. Expression levels greater than 30-fold higher are obtained using the ExpiCHO kit as directed versus substitution of PEI as a transfection reagent, while also using 50% less DNA.

For transfection of ExpiCHO cells, can I really use only 0.5-0.8 µg/mL plasmid DNA? Can lower DNA doses benefit hard-to-express proteins?

ExpiFectamine CHO Transfection Reagent is a highly efficient transfection reagent and as such significantly lower levels of plasmid DNA not only can, but in most instances should, be used for expression runs. Higher levels of DNA can be more stressful to the cells. The volume of ExpiFectamine CHO Reagent dictated in the kit protocol will account for using plasmid DNA in the range of 0.5-0.8µg/mL; if using less DNA than this, the amount of ExpiFectamine CHO Reagent should be reduced proportionately for best results. We recommend that you use 0.6-0.8 µg/mL for most proteins. For some proteins that are aggregate-prone or otherwise difficult to express, lower DNA doses may benefit expression.

What if I cannot add the diluted ExpiFectamine CHO Transfection Reagent to the diluted plasmid DNA immediately?

If it is not possible to immediately add diluted ExpiFectamine CHO Transfection Reagent to diluted plasmid DNA, we recommend diluting your plasmid DNA in the total complexation volume that would be used according to the kit protocol (i.e., the total volume of OptiPro Reagent that would normally be used for diluting both the ExpiFectamine CHO Reagent and the plasmid DNA) and then adding non-diluted ExpiFectamine CHO Reagent directly to the diluted plasmid DNA and then mixing by gentle pipetting 1-2 times and/or inversion. This method is useful for automation or small-scale transfections where it is impossible or undesirable to add the ExpiFectamine CHO Reagent to plasmid DNA immediately after dilution.

I am new to the ExpiCHO system and would like additional information to help me get started? Are there any online resources to assist me?

Yes. Please visit our website (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-expression/mammalian-protein-expression/transient-mammalian-protein-expression/expicho-expression-system.html?SID=fr-expichosystem-main) where you will find short videos (including https://www.youtube.com/watch?v=l2emhdSx0e4) showing exactly how ExpiCHO experiments are performed in our laboratories. This is an excellent resource for getting started or for troubleshooting.

Sometimes in the high-titer or max-titer ExpiCHO Expression System protocols, I see a drop in viability on or about day 7-8 posttransfection. How can I fix this?

This drop in viability tends to indicate that some aspect of the cell culture conditions are not optimal during the expression run. If this is observed, the shaking speed of the flasks should be verified to be within protocol specifications; the volume and size of the flask should be appropriate; and care should be taken when handling the ExpiCHO-S Cells ahead of the expression run to ensure that cells are not stressed by vigorous mixing, etc.

In some instances, flask-to-flask variability has been observed using the high- and max-titer protocols at the 125 mL flask scale. Here, it is recommended to increase the shaking speed to 130 rpm for 25 mm shakers and 140 rpm for 19 mm shakers.

Alternatively, we have found that baffled flasks work well for the 125 mL scale to correct any flask-to-flask variability—in such an instance, to account for the baffles, the volume of cells to be transfected should be increased from 25 mL to 35-40 mL to allow for optimal flow over the baffles, and the volumes of other reagents should be scaled accordingly.

Lower transfection volumes (i.e., 30-35 mL) can also be used with baffled flasks; however, shaking speeds must be reduced slightly to account for the baffles (i.e., 115 rpm for 19 mm shakers and 110 rpm for 25 mm shakers). We have not found baffled flasks to be necessary or beneficial at other flask sizes.

Can an ExpiCHO expression run be scaled down to smaller volumes than that in shake flasks?

Yes. For expression in 24- and 96-deep well blocks and 50 mL mini bioreactor tubes, please refer to the online protocol (https://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/Protein%20Bio%20-%20Thermo/pdf/ExpiCHO-Protocols-Deep-Well-Blocks-Mini-Bioreactor-Tubes.pdf)

Sometimes the ExpiCHO cell supernatant is difficult to filter during clarification. How can I overcome this?

Because the ExpiCHO system uses components that differ from that of HEK 293-based expression systems such as the Gibco Expi293 system, the supernatant may be more difficult to process through standard bottle top filters in some instances. To remedy this, we recommend centrifuging the supernatant first at ~5,000 x g for 30 minutes followed by filtration using a 0.22 µm filter. Alternative filters (such as depth filters) provide superior filtration for supernatants, especially at larger scales, as these filters are specifically designed to handle supernatant from CHO-derived expressions.

For the ExpiCHO Expression System protocol, what day should I harvest my supernatant?

The time of harvest is highly dependent upon the nature of your protein. For stable proteins such as antibodies, we generally recommend harvesting on day 7-8 posttransfection (using the standard protocol), on day 9-10 (using the high-titer protocol, and on day 11-12 (using the max-titer protocol. Cell viability should still be high at this time, ideally 70-80% or greater. Intracellular proteins may require earlier harvesting, between days 4-7. For proteins that may be susceptible to degradation, a time course of harvesting should be considered to identify the optimal takedown time. In all instances, maintaining high cell viability at the time of harvest is ideal for both protein quality and purification.

Are there any special instructions for the high-titer and max-titer protocols in the ExpiCHO Expression System?

When shifting the temperature for the high-titer and max-titer protocols, it is recommended that a dedicated 32 degrees C incubator be used, as simply changing the temperature of a 37 degrees C incubator to 32 degrees C may take a prolonged period of time to cool down and may limit the effectiveness of the temperature shift. Cooling the incubator by opening the door may result in contamination.

When should the ExpiCHO Feed and ExpiFectamine CHO Enhancer be added to the cultures?

ExpiCHO Feed and ExpiFectamine CHO Enhancer should be added 18-22 hours posttransfection for best results. These solutions may be added to the flasks without prewarming. ExpiCHO Feed and ExpiFectamine CHO Enhancer may also be premixed ahead of addition to flasks to reduce the number of steps required. If using the standard protocol at 37 degrees C, it is recommended to add the feed and enhancer closer to the 18-hour timepoint.

Which vector do you recommend using with the ExpiCHO Expression System?

We recommend using the pcDNA3.4 TOPO TA vector (Cat. No. A14697) for optimal expression. This vector contains the native, full-length cytomegalovirus (CMV) promoter and a woodchuck posttranscriptional regulatory element (WPRE) downstream of the cloning site. Other CMV-based vectors, however, have been shown to express similarly to the pcDNA3.4 TOPO TA vector, thus any CMV-driven vector is a good choice for the ExpiCHO system.

Can I use a different transfection reagent besides ExpiFectamineCHO Transfection Reagent for the transfection of ExpiCHO-S Cells?

The ExpiFectamine CHO Transfection Reagent, ExpiFectamine CHO Enhancer, and ExpiCHO Feed are optimized to work together to provide maximal protein expression levels and are provided as a single kit for convenience. The ExpiFectamineCHO Transfection Reagent provides high transfection efficiency of high-density cultures, superior to any other transfection reagent. Expression levels greater than 30-fold higher are obtained using the ExpiCHO kit as directed versus substitution of polythyleneimine (PEI) for the transfection reagent, while using 50% less DNA in both methods.

Are there any tips regarding the optimal proportion of light- and heavy-chain—encoding plasmids for optimal antibody expression when using the ExpiCHO Expression System?

For the majority of antibodies tested, a 1:1 ratio has been found to be ideal for the ExpiCHO system. The optimal heavy- to light-chain plasmid ratio is dependent upon the rate at which the two chains are expressed when cotransfected into the same cell, and may be antibody specific in some instances. We recommend starting with this ratio and then modifying as necessary for specific molecules. We recommend cloning the heavy- and light-chain subunits separately into the Invitrogen pcDNA3.4 TOPO vector initially, and then optimizing the ratios of the two plasmids. We have also tested single plasmids incorporating both heavy- and light-chain genes with comparable results, so in the end, the answer will come down to user experience and preference.

Can I really use only 0.5-0.8 µg/mL plasmid DNA with ExpiFectamine CHO reagent? Can lower DNA levels be used for hard-to-express proteins?

ExpiFectamine CHO is highly efficient for reagent transfection, enabling use of significantly lower levels of plasmid DNA for expression runs. Higher levels of DNA can be more stressful to the cells. The volume of ExpiFectamineCHO reagent specified in the kit protocol will account for using plasmid DNA in the range from 0.5 to 0.8 µg/mL; if using less DNA than this, the amount of ExpiFectamineCHO reagent should be reduced proportionately for best results. It is recommended to use 0.6-0.8 µg/mL for most proteins. For some proteins that are aggregate-prone or otherwise difficult to express, lower DNA levels may benefit expression.

What if I cannot add the diluted ExpiFectamine CHO reagent to the diluted plasmid DNA immediately?

If it is not possible to immediately add diluted ExpiFectamine CHO reagent to diluted plasmid DNA, we recommend diluting your plasmid DNA in the total complexation volume that would be used according to the kit protocol (i.e., the total volume of OptiPro media that would normally be used for diluting both the ExpiFectamine CHO reagent and the plasmid DNA) and then adding non-diluted ExpiFectamine CHO reagent directly to the diluted plasmid DNA. Mix by gentle pipetting 1-2 times and/or inversion. This method is useful for automation or small-scale transfections, where it is impossible or undesirable to add the ExpiFectamine CHO reagent to plasmid DNA immediately after dilution.

How can I ensure the best complexation of ExpiFectamine CHO reagent and my plasmid DNA?

For best results, it is recommended to perform DNA complexation in the following manner:

1. Dilute plasmid DNA into cold Gibco OptiPro medium.
2. At the time of use, dilute the ExpiFectamine CHO reagent with cold OptiPro medium and then immediately add to the diluted plasmid DNA.
3. Mix by gentle pipetting 1-2 times and/or inversion; do not vortex or pipet vigorously.
4. Complexation time should take 30 seconds - 5 minutes.

Avoid elongated hold times for the diluted ExpiFectamine CHO reagent or the ExpiFectamine CHO reagent-plasmid DNA complexation reaction.

What density should the ExpiCHO-S Cells achieve at the time of transfection?

ExpiCHO-S Cells should be subcultured at a density of 3-4 x 10E6 cells/mL one day prior to transfection to obtain a cell density of approximately 7-10 x 10E6 cells/mL on the day of transfection. These cells should be diluted to a final density of 6 x 10E6 cells/mL with fresh media and gently swirled to mix prior to transfection. Discard any remaining cells; do not reuse high-density cells for seeding of maintenance flasks.

Can I substitute other media for ExpiCHO medium for culturing ExpiCHO-S Cells?

No. The ExpiCHO Expression System is able to achieve very high titers due to the way the components of the system have been optimized to work together for maximal protein expression. ExpiCHO medium is a transfection-compatible, high-density growth medium specifically matched to Gibco ExpiCHO Feed and ExpiFectamine CHO Enhancer. Other media are not compatible with the ExpiCHO system and may inhibit protein expression altogether.

How can I ensure that my ExpiCHO-S Cells are growing optimally?

As a quick check, ExpiCHO-S Cells can be seeded at 0.3 x 10E6 cells/mL in 30 mL of ExpiCHO medium in a 125 mL non-baffled flask, and the viability and viable cell density checked on days 5, 6, 7 postseeding. Typically, ExpiCHO-S Cells will reach maximal density around day 6 postseeding and should attain a density in the range of 20 x 10E6 cells/mL, and then will die off on day 7 and beyond. Determining the final viable cell density will be dependent upon the method used to count cells. Significant variability can be observed from different counting methods. If cells are exhibiting significantly different growth profiles, optimization of culture conditions should be performed. In such instances, it is typically useful to test multiple different shaking speeds simultaneously to determine which speed provides optimal cell growth and then start with this speed for your protein expression runs.

What should I do if my ExpiCHO-S Cells overgrow the 4-6 x 10E6 cells/mL target density ahead of subculturing?

If cells significantly overgrow the target of 4-6 x 10E6 cells/mL (i.e., growing to 8-10 x 10E6 cells/mL), simply subculture the cells to a higher cell density than normal (i.e., seed new flasks at ~0.5 x 10E6 cells/mL) to allow the cells to recover. Since ExpiCHO-S Cells belong to a high-density cell line, it is not recommended to subculture the cells if they have not yet reached log phase of growth at 4-6 x 10E6 cells/mL, as this can impair growth over time.

How soon after thawing can I use my ExpiCHO-S Cells and how many passages are they good for?

ExpiCHO-S Cells should be passaged at least twice post-thaw and be growing within the ranges specified in the ExpiCHO system manual, prior to transfection. Cells should maintain consistent performance for at least 20 passages if maintained in accordance with the cell culture maintenance guidelines in the manual.

What are the best practices for handling ExpiCHO-S Cells?

ExpiCHO-S Cells do not reach log phase of growth until approximately 3 x 10E6 cells/mL cells, thus ExpiCHO-S Cells should be allowed to attain a density of 4-6 x 10E6 cells/mL at the time of subculturing to ensure the cells have reached log phase of growth. As cells approach 6 x 10E6 cells/mL, some very small cell clumps may be visible; do not try to break up the clumps, simply let them settle and remove the suspended cell solution for subculturing. For all cell manipulations, simply swirl flasks to resuspend the cells. Do not shake or pipet the cells vigorously to mix, as this can lead to decreased performance, especially just prior to transfection, when cells have attained very high densities.

I've followed all of the recommendations for thawing my ExpiCHO-S Cells, but they are not growing post-thaw. What can I do?

In instances where ExpiCHO-S Cells are thawed and do not start to grow as noted above—attaining only relatively low densities in culture—one common solution is to verify the temperature of the cultures to ensure that the equipment settings are not generating too much heat. CHO cells are very tolerant to lower temperatures, but are more sensitive to elevated temperatures compared to HEK293 cells. Incubators, even when set to 37 degrees C, in conjunction with the heat generated from the shakers, can elevate the temperature in the culture flasks above optimal for CHO cells.

The optimal temperature of the media in the flask for ExpiCHO-S Cells should be around ~36.5 degrees C. If overheating is suspected, reduce the temperature of the incubator to reach optimal operating temperatures in the flasks. In such instances, it is best practice to thaw a new vial of cells rather than attempt to recover cells from elevated temperature conditions. Also, non-baffled flasks are recommended for use at all scales for both routine subculturing and protein expression runs.

What are the normal growth characteristics of ExpiCHO-S Cells?

Within 1-2 passages post-thaw, ExpiCHO-S Cells should be growing with a doubling time of approximately 18 hours. There should be minimal cell clumping in the flasks, with only small clumps visible when the cells approach higher cell densities (i.e., ~6 x 10E6 cells/mL). When cells are cultured at 0.2-0.3 x 10E6 cells/mL, or 0.1-0.2 x 10E6 cells/mL, viable cell density should be approximately 4-6 x 10E6 cells/mL within 3 or 4 days, respectively. If cells are not growing within these approximate ranges, cell culture conditions will require further optimization.

How should the Gibco ExpiCHO-S Cells be handled upon receipt?

Upon receipt of the cells on dry ice, it is best to either thaw the cells immediately or place the vials into liquid nitrogen storage for ~72 hours to allow cells to acclimate until the time of thaw; do not store cells at -80 degrees C. Once thawed and transferred into prewarmed media in a vented, non-baffled shake flask, cells should be incubated at 37 degrees C with 8% CO2 on a shaker platform set to 120 ±5 rpm for a shaker with a 25 mm orbital diameter or 125 ±5 rpm for a 19 mm orbital diameter. Cells should have high viability at the time of thaw and should recover quickly post-thaw, reaching their normal 18-hour doubling time within 1-2 passages.

Cells that are very clumpy or stringy in appearance or do not recover into a normal growth pattern within 1-2 passages may have been compromised during shipping or receiving, and shouldn't be used.

I'm new to the Gibco ExpiCHO Expression System and would like additional information to help me get started. Are there any online resources to assist me?

Yes. Please visit the ExpiCHO page at thermofisher.com/expicho where you will find short videos showing exactly how expression with the ExpiCHO system is performed in our laboratories. This is an excellent resource for getting started or for troubleshooting.

I left my DNA:Expifectamine CHO complexes sitting longer than 5 minutes. Will they be okay?

The optimal complexation time is 5 minutes. We have observed a small drop in protein yield (approximately 20% reduction) if complexes sit up to 10 minutes; for 20 minutes or longer yield will be drastically reduced (>50% reduction in yield).

I am getting lower yield than I expected with the ExpiCHO expression system. What should I do to improve my results?

Yield can vary greatly from protein to protein. We strongly recommend expressing the rabbit antibody positive control (Cat. No. A14662) to determine if the low yield is due to a low expressing protein, a problem with the system components, or transfection and culturing conditions. If you are not achieving the expected yield with the rabbit IgG positive control, we recommend checking the following:

- Ensure that cells are >95% viable during normal passaging and at time of transfection
- Have a doubling time of approximately 17 h
- Recover cells rapidly post-thaw (within 3-4 days post thaw); if not, verify you are using the culturing guidelines provided in the manual and thaw a new vial of cells if necessary.
- We recommend gentle swirling at all handling steps for ExpiCHO-S cells (avoid vigorous swirling, shaking, and pipetting up and down). Verify that your shake speed is about 110-125 rpm for 25 mm throw shakers and about 125 for 19 mm throw shakers. Test a flask with containing water and a thermometer to determine whether the shaker is putting off excess heat; reduce the incubator temperature setting as necessary to achieve a final temperature of 37 degrees C for your cultures. All of our shake speed recommendations are provided for Corning non-baffled flasks; if you are using a different culture vessel, additional shake speed optimization may be required.

I notice my ExpiCHO-S cells are clumping. What should I do?

Some mild cell clumping is normal during ExpiCHO-S cell passaging and is okay as long as the cells have >95% viability. At each passage you may allow the clumps to settle to the bottom by letting the flask rest for 30 sec to 1 min, then passage the suspended cells. If the cells have a lot of clumping paired with viability < 95%, this indicates a problem either with the culture conditions or the cells themselves. We recommend verifying that the cells are: cultured according to the recommendations in the manual, below passage 20, and free of contamination. Thaw a new vial of cells as necessary.

What method do you recommend for quantifying the rabbit Antibody-Expressing Positive Control Vector (Cat. No. A14662)?

The formation of intact IgG molecules may be quantitated using a sandwich ELISA designed to capture and detect rabbit IgG. Besides the rabbit IgG positive control, reagents, and consumables that are included in the kit, you will also need purified rabbit IgG to be used as a standard, F(ab')2 goat anti-rabbit IgG HRP conjugate (Cat. No. A10547), Protein A-coated plates (Cat. No. 15130 for clear plates used in colorimetric detection), TMB colorimetric substrate (Cat. No. 34021), SuperBlock (TBS) Blocking Buffer (Cat. No. 37581), and PBS or TBS buffer for washes. There is an example procedure in our Protein A-coated plates manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0011310_Pierce_ProteinA_G_AG_Coat_96Well_UG.pdf). Please note, our R&D scientists determine titer values from crude cell culture supernatants using a Pall Life Sciences FortéBio Octet instrument equipped with a protein A biosensor.

What viability is typical for ExpiCHO-S cells, and what method do you recommend for monitoring cell viability of ExpiCHO-S cells?

For both normal passaging and at the time of transfection for ExpiCHO-S cells, greater than 95% viability is critical for best results with the ExpiCHO expression system. We recommend for monitoring cell viability to use a trypan blue exclusion method (either manual or automated using a Vi-CELL Cell Viability Analyzer or Cedex Analyzer). Trypan blue stain is available for purchase (Cat. No. 15250061).

How does the protein expression yield of the ExpiCHO expression system compare to FreeStyle CHO or Expi293 expression systems?

With several test proteins in the ExpiCHO-S expression system, we see approximately 25-160 fold higher expression than FreeStyle CHO expression system and approximately 2-4 fold higher expression than Expi293 expression system. Please see Figure 1 on the ExpiCHO web page at the following link for the data: http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-expression/mammalian-protein-expression/transient-mammalian-protein-expression/expicho-expression-system.htmL?icid=npiGP-MJ-NP01-expicho-expression-system-20150908

Can ExpiCHO-S Cells be used to create stable clones?

Our ExpiCHO-S Cells are derived from our cGMP banked CHO-S cells (Cat. No. A1155701), thus stable CHO-S selection methods are applicable to these cells. We recommend you follow the protocol outlined in the User Bulletin for the Creation and Scale up of a stable cell ine using ExpiCHO Products (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017764_CreatScaleup_StableCellExpiCHO_UB.pdf)

Can I substitute a different medium for ExpiCHO Expression Medium in my Expifectamine CHO transfection?

We do not recommend substituting another medium for ExpiCHO Expression Medium because it likely cannot support the same viable density as ExpiCHO Expression Medium, and may also contain components that inhibit the transfection using ExpiFectamine CHO.

I don't have ExpiCHO-S cells. Can Expifectamine CHO transfection reagent and enhancer be used with other CHO cell lines than the ExpiCHO-S cell line?

Yes, you may be able to adapt your CHO cells into ExpiCHO medium. Long-term adaptation in ExpiCHO medium may increase productivity of your CHO cells, and should sustain high-density growth. However there is no guarantee that CHO lines other than Expi CHO-S cell line will achieve the same levels of expression as the ExpiCHO-S cells. In limited testing, we have found other CHO subclones to be less easy to transfect than ExpiCHO-S cells in large-scale suspension format.

Do I need to change the media after the ExpiFectamine CHO transfection?

No. The ExpiCHO Expression System is designed to run without media exchanges. There is no need to remove transfection complexes or to change growth medium following transfection, however there are an enhancer addition and 1-2 optional feed additions for improved yield.