Learn More
Gibco™ Expi293™ PRO Transfection Kit
The Expi293 PRO Transfection Kit enables high-efficiency delivery of DNA into HEK293 cells, supporting fast, scalable protein expression. Optimized for use with Expi293F cells, it delivers robust performance across a broad range of protein types.
$1,487.70 - $8,148.60
Specifications
| Cell Type | HEK293 |
|---|---|
| Format | Kit |
| Sample Type | Plasmid DNA |
| Promoter | pcDNA™ 3.4 |
| Transfection Technique | Lipid-based Transfection, Transient Transfection |
| Catalog Number | Mfr. No. | Sufficient For | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sufficient For | Price | Quantity | |||||
|
A40002926
|
Gibco™
A40002926 |
For 1 L transfection |
N/A
|
|
|||||
|
A40002929
|
Gibco™
A40002929 |
For 6 L transfection |
N/A
|
|
|||||
|
A40002927
|
Gibco™
A40002927 |
For 10 L transfection |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
A40002930
|
Gibco™
A40002930 |
For 50 L transfection |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The Expi293 PRO Transfection Kit is a core component of the Expi293 PRO Expression System. It is designed for transient transfection of high density cultures of HEK 293 cells. The high-efficiency, cationic, lipid-based transfection reagent and transfection enhancers are designed to power the highest possible level of protein expression from Expi293 PRO cells cultured in Expi293 PRO Expression Medium.
- Designed specifically for transfection of high-density suspension cell culture, with complimentary feed and enhancers that boost transfection performance and protein expression
- Achieves protein yields 2- to 10-fold higher than other transfection reagents used on high-density Expi293 PRO cell cultures
- Provides robust and reproducible transfection results, giving users greater confidence in their results
- Scale transfections for culture volumes of less than 1 mL to greater than 10 liters, while maintaining equivalent volumetric protein yields
The Expi293 PRO Transfection Kit is specifically designed to work with Expi293 PRO cells in the Expi29 PRO Expression System. The transfection kit consists of the cationic lipid-based Expi293 PRO transfection reagent and Expi293 PRO enhancer and feed, which boost transfection, cell viability, and protein expression.
Specifications
| HEK293 | |
| Plasmid DNA | |
| Lipid-based Transfection, Transient Transfection | |
| Transfection Kit |
| Kit | |
| pcDNA™ 3.4 | |
| Expi293 PRO | |
| Cell Culture Grade |
For Research Use Only. Not for use in diagnostic procedures.