missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ MembranePro™ Expression System

Catalog No. p-7075865
Encompass
Change view
Click to view available options
Quantity:
10 Reactions
100 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25869 10 Reactions
A25870 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25869 Supplier Gibco™ Supplier No. A25869
Only null left

Expi293™ MembranePro™ Expression System

For researchers studying G protein–coupled receptors (GPCRs) and other membrane proteins, the Expi293™ MembranePro™ Expression System delivers enriched, functional membrane proteins with high efficiency, scalability and reliability for a wide variety of low or high throughput applications.

Features of the Expi293™ MembranePro™ Expression System include:
  • Efficient—more than 20-fold increase in membrane protein yield compared to the adherent culture system
  • Scalable—easy scale-up without the need for multiple flasks
  • Easy to use—no enzymatic or mechanical dissociation required

    In addition, benefits of MembranePro™ technology include:
  • Proteins are displayed on human (or other mammalian) cell membranes
  • Cellular quality control and mammalian post-translational processing helps produce functional protein
  • Proteins bud off as lipoparticles and are easily collected from the culture medium
  • Lipoparticles are enriched with expressed receptors

    With the less labor-intensive Expi293™ MembranePro™ Expression System protocol, you will:
    1. Clone your gene of interest using a TOPO™ vector
    2. Transfect a susupension culture of Expi293F™ cells using Expifectamine™ 293 transfection reagent
    3. Harvest lipoparticles 48 hrs after addition of enhancers using MembranePro™ Precipitation Mix
    4. Store lipoparticles at -80 C or proceed to downstream assays

    Note: Exi293F™ Cells (A14527) and a pEF6 V5-His TOPO™ Expression Kit (K9610-20) are required for use of this system and must be purchased separately when needed.
  • Specifications

    Type Expression System
    Sufficient For 10 Reactions
    Delivery Type Virus-Like Particles (VLPs)
    Quantity 10 Reactions
    Content And Storage • MembranePro™ Reagent, 1 x 0.3 mg (1 mg/mL), store at -5 to -30°C
    • MembranePro™ Precipitation Mix, 1 x 75 mL, store at room temperature
    • 1 ExpiFectamine™ 293 transfection kit (containing reagent and enhancers) for 1 L of culture, store at 2–8°C
    • Opti-MEM™ I Reduced Serum Medium, 1 x 100 mL, store at 2–8°C
    • Expi293™ Expression Medium, 1 x 1 L, store in the dark at 2–8°C
    Cell Line Expi293F™
    Product Line Expi293, MembranePro
    For Use With (Application) Membrane Protein Expression
    Promoter EF-1α

    For Research Use Only. Not for use in diagnostic procedures.