Learn More
Gibco™ Expi293™ Expression Medium
Expi293 Expression Medium designed for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells.
Supplier: Gibco™ A1435103
Description
Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F cells in the Expi293 Expression System for scalable transient protein expression.
- Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
- Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine transfection reagent
- Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
- Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10 L in disposable bioreactors
- Does not contain phenol red
Expi293 Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.
Specifications
| Expi293F™ | |
| Liquid | |
| GlutaMAX | |
| Expi293 | |
| One 10L Universal Bag Store in the dark at 2-8°C. |
| Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
| Expression Medium | |
| No Phenol Red | |
| 10 L |
For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.