missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ExoSAP-IT Express PCR Product Cleanup with Tracking Dye DFS Item

Our fastest PCR clean-up method - just 5 minutes to superior sequencing results. Now available with optional tracking dye. It is necessary to remove excess primers and unincorporated nucleotides from the PCR reaction prior to sequencing.

$1020.00 - $3950.00

Specifications

Content And Storage Shipped on dry ice. Store at -20°C.
Product Line ExoSAP-IT
Protein Form Recombinant
Research Category Genetic Diseases
Shipping Condition Dry Ice
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity  
Catalog Number Mfr. No. No. of Reactions Price Quantity  
A35004 DFS Item
View Documents
Applied Biosystems™
A35004
500 Reactions
Each for $1,020.00
Only null left
 
A35005 DFS Item
View Documents
Applied Biosystems™
A35005
1920 reactions
Each for $3,950.00
Only null left
 
Description

Description

  • 5 minute protocol: Fastest turnaround time to results
  • One-tube, one-step PCR clean-up: Add ExoSAP-IT Express directly to PCR product
  • Novel enzyme technology: Irreversibly inactivated in just one minute at 80°C
  • Conserve PCR samples: 100% recovery of PCR products, regardless of amplicon length
  • Eliminate spin columns or beads: Significantly decreases time and errors while increasing yield
  • Compatible: No interference in downstream applications
  • Scalable: Treat a wide range of volumes
  • Multiple formats: Single tube, 8 tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
  • Go green: Generate less waste with a single-tube solution to PCR clean-up
  • Optional tracking dye: Never lose track of reagent addition during pipetting
Specifications

Specifications

Shipped on dry ice. Store at -20°C.
Recombinant
Dry Ice
ExoSAP-IT
Genetic Diseases
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.