Learn More
Applied Biosystems™ ExoSAP-IT Express PCR Product Cleanup with Tracking Dye
Description
- 5 minute protocol: Fastest turnaround time to results
- One-tube, one-step PCR clean-up: Add ExoSAP-IT Express directly to PCR product
- Novel enzyme technology: Irreversibly inactivated in just one minute at 80°C
- Conserve PCR samples: 100% recovery of PCR products, regardless of amplicon length
- Eliminate spin columns or beads: Significantly decreases time and errors while increasing yield
- Compatible: No interference in downstream applications
- Scalable: Treat a wide range of volumes
- Multiple formats: Single tube, 8 tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
- Go green: Generate less waste with a single-tube solution to PCR clean-up
- Optional tracking dye: Never lose track of reagent addition during pipetting
Specifications
Specifications
| Content And Storage | Shipped on dry ice. Store at -20°C. |
| Format | 8 Tube Strip, 96-well Plate, Tube |
| For Use With (Application) | PCR Purification |
| No. of Reactions | 1920 reactions |
| Product Line | ExoSAP-IT |
| Protein Form | Recombinant |
| Quantity | 1,920 reactions (96-well plate, 40 μL per we |
| Research Category | Genetic Diseases |
| Shipping Condition | Dry Ice |
| Test Time | 5 min. |
Frequently Asked Questions (FAQs)
The HT ExoSAP-IT Fast High-Throughput PCR Product Cleanup reagent is designed for automated handlers as it has a lower viscosity than standard ExoSAP-IT PCR Product Cleanup reagent, which contains some glycerol.
The Quant-iT PicoGreen dsDNA Assay Kit is recommended for quantifying PCR products that have been cleaned with ExoSAP-IT reagent. ExoSAP-IT PCR cleanup results in 100% recovery regardless of amplicon length, and if the PCR product was quantified prior to ExoSAP-IT cleanup, additional quantification is not necessary. We do not recommend using a spectrophotometer as degraded products will be included in the quantification.
For use with reverse transcription, ExoSAP-IT does not degrade the DNA-RNA hybrid. However, it will degrade single-stranded cDNA if a 3' -OH end is present.
The tracking dyes used in the DreamTaq Hot Start Green PCR Master Mix have not been shown to interfere with functionality of the ExoSAP-IT cleanup reagents.
The proofreading function of HiFi DNA polymerases can cause partial degradation of PCR products during extended incubation at 80°C. ExoSAP-IT Express reagent is recommended for HiFi PCR cleanup because it only requires 1 minute at 80°C for inactivation.
For Research Use Only. Not for use in diagnostic procedures.