Learn More
Applied Biosystems™ EXOSAPIT EXPRESS PCR PRO 40 UL
Catalog No. 501124990
Change view
Click to view available options
:
20 reactions
Catalog No.
501124990 20 reactions
1 options
Catalog No. 501124990 Supplier Applied Biosystems™ Supplier No. LS7500140UL
Only null left
Removes excess primers and dNTPs. Does not interfere with downstream applications.
  • Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
  • One-tube, one-step PCR cleanup
  • Five-minute protocol
  • Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows