missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Evodiamine

Catalog No. AAJ62771MD
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62771MD 250 mg
AAJ62771MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62771MD Supplier Thermo Scientific Chemicals Supplier No. J62771MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 518-17-2 | C19H17N3O, C19H17N3O | 303.37 g/mol

A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 518-17-2
Molecular Formula C19H17N3O

Specifications

Quantity 250 mg
Description Off-white to pale yellow
Content And Storage -30°C to -10°C
For Use With (Application) A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
Formula Weight 303.40
Product Type Evodiamine
Form Solid

RUO – Research Use Only