missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Etodolac

Catalog No. AC465720050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465720050 5 g
AC465720250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465720050 Supplier Thermo Scientific Chemicals Supplier No. 465720050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41340-25-4
Molecular Formula C17H21NO3

Specifications

Quantity 5 g
Melting Point 150.0°C to 152.0°C
Formula Weight 287.35
InChI Key NNYBQONXHNTVIJ-UHFFFAOYNA-N
SMILES CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1
IUPAC Name 2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
Molecular Weight (g/mol) 287.36
Chemical Name or Material Etodolac
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes serious eye irritation.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only