missing translation for 'onlineSavingsMsg'
Learn More

Ethynyl p-Tolyl Sulfone 98.0+%, TCI America™

Catalog No. E04661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04661G 1 g
E04665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04661G Supplier TCI America Supplier No. E04661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13894-21-8
Molecular Formula C9H8O2S
Molecular Weight (g/mol) 180.221
MDL Number MFCD00191647
InChI Key FTHKWIMQNXVEHW-UHFFFAOYSA-N
Synonym p-Toluenesulfonylacetylene, Tosylacetylene
PubChem CID 4463273
IUPAC Name 1-ethynylsulfonyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C#C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 180.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethynyl p-Tolyl Sulfone