missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. E0382500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0382500G 500 g
E038225G 25 g
E0382100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0382500G Supplier TCI America Supplier No. E0382500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1530-32-1
Molecular Formula C20H20BrP
Molecular Weight (g/mol) 371.26
MDL Number MFCD00011838
InChI Key JHYNXXDQQHTCHJ-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
PubChem CID 73727
IUPAC Name ethyltriphenylphosphanium bromide
SMILES [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 500 g
UN Number 2811
Formula Weight 371.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyltriphenylphosphonium Bromide