missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium bromide, 98%

CAS: 1530-32-1 | C20H20BrP | 371.26 g/mol

$68.88 - $68.88

Chemical Identifiers

CAS 1530-32-1
Molecular Formula C20H20BrP
Molecular Weight (g/mol) 371.26
MDL Number MFCD00011838
InChI Key JHYNXXDQQHTCHJ-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
PubChem CID 73727
IUPAC Name ethyl(triphenyl)phosphanium;bromide
SMILES [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119040250
SDS View Documents
Thermo Scientific Chemicals
119040250
25 g Glass bottle
Each for $68.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

1530-32-1
371.26
JHYNXXDQQHTCHJ-UHFFFAOYSA-M
73727
[Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C20H20BrP
MFCD00011838
ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
ethyl(triphenyl)phosphanium;bromide

Specifications

1530-32-1
100.0
White
1% max.
97.5% min. (on dry substance) (Argentometry)
C20H20BrP
MFCD00011838
ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
JHYNXXDQQHTCHJ-UHFFFAOYSA-M
ethyl(triphenyl)phosphanium;bromide
73727
98%
Ethyltriphenylphosphonium bromide, 99+%
97.5
206.5°C to 208.5°C
>200°C
Authentic
Glass bottle
C2H5P(C6H5)3Br
25 g
Solubility in water: 120g/L (23°C). Other solubilities: soluble in chloroform,fairly soluble in lower alcohols,slightly soluble in arom. hydrocarbons
[Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
371.26
371.26
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/

GHS Signal Word: Danger

EINECSNumber : 216-223-3

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only