missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium bromide, 98%

Catalog No. AC119045000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119045000 500 g Glass bottle
AC119040250 25 g Glass bottle
AC119041000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC119045000 Supplier Thermo Scientific Chemicals Supplier No. 119045000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1530-32-1 | C20H20BrP | 371.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1530-32-1
Molecular Formula C20H20BrP
Molecular Weight (g/mol) 371.26
MDL Number MFCD00011838
InChI Key JHYNXXDQQHTCHJ-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
PubChem CID 73727
IUPAC Name ethyl(triphenyl)phosphanium;bromide
SMILES [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 206.5°C to 208.5°C
Color White
Flash Point >200°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula C2H5P(C6H5)3Br
Quantity 500 g
Solubility Information Solubility in water: 120g/L (23°C). Other solubilities: soluble in chloroform,fairly soluble in lower alcohols,slightly soluble in arom. hydrocarbons
Formula Weight 371.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyltriphenylphosphonium bromide, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/
EINECSNumber 216-223-3
TSCA TSCA

RUO – Research Use Only