Learn More
(Ethylthio)acetic acid, 97%
CAS: 627-04-3 | C4H8O2S | 120.166 g/mol
$77.47 - $229.81
Chemical Identifiers
| CAS | 627-04-3 |
|---|---|
| Molecular Formula | C4H8O2S |
| Molecular Weight (g/mol) | 120.166 |
| MDL Number | MFCD00021769 |
| InChI Key | VJIKFWJCVWFZIN-UHFFFAOYSA-N |
| Synonym | ethylthio acetic acid, acetic acid, ethylthio, ethylthioacetic acid, ethylmercapto acetic acid, carboxymethyl ethyl sulfide, s-carboxymethyl ethyl mercaptan, 2-ethylthio acetic acid, unii-m3d5y724hz, acetic acid, 2-ethylthio, ethylmercaptoacetic acid |
| PubChem CID | 69387 |
| IUPAC Name | 2-ethylsulfanylacetic acid |
| SMILES | CCSCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL0474306
|
N/A | 5 g |
Each for $77.47
|
|
|||||
|
AAL0474314
|
N/A | 25 g |
Each for $229.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C4H8O2S | |
| MFCD00021769 | |
| ethylthio acetic acid, acetic acid, ethylthio, ethylthioacetic acid, ethylmercapto acetic acid, carboxymethyl ethyl sulfide, s-carboxymethyl ethyl mercaptan, 2-ethylthio acetic acid, unii-m3d5y724hz, acetic acid, 2-ethylthio, ethylmercaptoacetic acid | |
| 2-ethylsulfanylacetic acid |
Specifications
| 627-04-3 | |
| C4H8O2S | |
| MFCD00021769 | |
| 1743038 | |
| VJIKFWJCVWFZIN-UHFFFAOYSA-N | |
| 2-ethylsulfanylacetic acid | |
| 69387 | |
| 97% |
| 117°C to 118°C (11 mmHg) | |
| 1.486 | |
| UN3265 | |
| ethylthio acetic acid, acetic acid, ethylthio, ethylthioacetic acid, ethylmercapto acetic acid, carboxymethyl ethyl sulfide, s-carboxymethyl ethyl mercaptan, 2-ethylthio acetic acid, unii-m3d5y724hz, acetic acid, 2-ethylthio, ethylmercaptoacetic acid | |
| CCSCC(=O)O | |
| 120.166 | |
| 120.18 | |
| (Ethylthio)acetic acid |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 210-979-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only