missing translation for 'onlineSavingsMsg'
Learn More

(Ethylthio)acetic acid, 97%

Catalog No. AAL0474314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0474314 25 g
AAL0474306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0474314 Supplier Thermo Scientific Chemicals Supplier No. L0474314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 627-04-3 | C4H8O2S | 120.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-04-3
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00021769
InChI Key VJIKFWJCVWFZIN-UHFFFAOYSA-N
Synonym ethylthio acetic acid, acetic acid, ethylthio, ethylthioacetic acid, ethylmercapto acetic acid, carboxymethyl ethyl sulfide, s-carboxymethyl ethyl mercaptan, 2-ethylthio acetic acid, unii-m3d5y724hz, acetic acid, 2-ethylthio, ethylmercaptoacetic acid
PubChem CID 69387
IUPAC Name 2-ethylsulfanylacetic acid
SMILES CCSCC(=O)O

Specifications

Boiling Point 117°C to 118°C (11 mmHg)
Refractive Index 1.486
Quantity 25 g
UN Number UN3265
Beilstein 1743038
Formula Weight 120.18
Percent Purity 97%
Chemical Name or Material (Ethylthio)acetic acid
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-979-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only