accessibility menu, dialog, popup

UserName

H3CSOHO

CAS RN 627-04-3

IUPAC Name: 2-(ethylsulfanyl)acetic acid
Synonyms: acetic acid, (ethylthio)-
Molecular Weight (g/mol): 120.17
Molecular Formula: C4H8O2S
InChi Key: VJIKFWJCVWFZIN-UHFFFAOYSA-N
SMILES: CCSCC(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)