Learn More
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™
CAS: 925-90-6 | C2H5BrMg | 133.27 g/mol
$122.33 - $122.33
Chemical Identifiers
| CAS | 925-90-6 |
|---|---|
| Molecular Formula | C2H5BrMg |
| Molecular Weight (g/mol) | 133.27 |
| MDL Number | MFCD00000043 |
| InChI Key | TWTWFMUQSOFTRN-UHFFFAOYSA-M |
| Synonym | ethylmagnesium bromide, grignard reagent, ethyl magnesium bromide, magnesium, bromoethyl, bromoethylmagnesium, bromoethylmagnesium, ethylmagnesium bromide solution, 3.0 m in diethyl ether, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesium bromid, ethylmagnesium bromid |
| PubChem CID | 101914 |
| SMILES | CC[Mg]Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC210471000
|
Thermo Scientific Chemicals
210471000 |
100 mL | AcroSeal™ Glass bottle |
Each for $122.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 925-90-6 | |
| 133.27 | |
| TWTWFMUQSOFTRN-UHFFFAOYSA-M | |
| 101914 |
| C2H5BrMg | |
| MFCD00000043 | |
| ethylmagnesium bromide, grignard reagent, ethyl magnesium bromide, magnesium, bromoethyl, bromoethylmagnesium, bromoethylmagnesium, ethylmagnesium bromide solution, 3.0 m in diethyl ether, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesium bromid, ethylmagnesium bromid | |
| CC[Mg]Br |
Specifications
| Black-Brown to Brown | |
| C2H5BrMg | |
| MFCD00000043 | |
| 02,205; 08,510; 11,574 | |
| ethylmagnesium bromide, grignard reagent, ethyl magnesium bromide, magnesium, bromoethyl, bromoethylmagnesium, bromoethylmagnesium, ethylmagnesium bromide solution, 3.0 m in diethyl ether, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesium bromid, ethylmagnesium bromid | |
| TWTWFMUQSOFTRN-UHFFFAOYSA-M | |
| magnesium;ethane;bromide | |
| 101914 | |
| AcroSeal™ Glass bottle | |
| 100 mL | |
| Solution |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Harmful if swallowed. May cause respiratory irritation. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
RUO – Research Use Only