missing translation for 'onlineSavingsMsg'
Learn More

Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals

$238.32 - $238.32

Chemical Identifiers

CAS 2146-37-4
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00019319
InChI Key VONKRKBGTZDZNV-UHFFFAOYSA-N
Synonym cyclopentane, ethylidene, ethyl idenecyclopentane, ethylidene-cyclopentane, cyclopentane,ethylidene, acmc-1cs2b, 1e-ethylidenecyclopentane, ethylidenecyclopentane, 90+%
PubChem CID 75068
IUPAC Name ethylidenecyclopentane
SMILES CC=C1CCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2020906
View Documents
Thermo Scientific Chemicals
B2020906
5 g
Each for $238.32
Only null left
 
Description

Description

It is used to make other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to make other organic chemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

2146-37-4
96.173
VONKRKBGTZDZNV-UHFFFAOYSA-N
75068
CC=C1CCCC1
C7H12
MFCD00019319
cyclopentane, ethylidene, ethyl idenecyclopentane, ethylidene-cyclopentane, cyclopentane,ethylidene, acmc-1cs2b, 1e-ethylidenecyclopentane, ethylidenecyclopentane, 90+%
ethylidenecyclopentane

Specifications

2146-37-4
0.8
C7H12
MFCD00019319
UN3295
Soluble in water.
CC=C1CCCC1
96.173
96.17
Ethylidenecyclopentane
-130°C
112°C to 114°C
1.45
5 g
cyclopentane, ethylidene, ethyl idenecyclopentane, ethylidene-cyclopentane, cyclopentane,ethylidene, acmc-1cs2b, 1e-ethylidenecyclopentane, ethylidenecyclopentane, 90+%
VONKRKBGTZDZNV-UHFFFAOYSA-N
ethylidenecyclopentane
75068
≥90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224
Extremely flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 218-413-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only