missing translation for 'onlineSavingsMsg'
Learn More

Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2020906

 View more versions of this product

Catalog No. AAB2020906


Only null left
Explore available promotions

Description

Description

It is used to make other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to make other organic chemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Ethylidenecyclopentane
-130°C
112°C to 114°C
1.45
5 g
cyclopentane, ethylidene, ethyl idenecyclopentane, ethylidene-cyclopentane, cyclopentane,ethylidene, acmc-1cs2b, 1e-ethylidenecyclopentane, ethylidenecyclopentane, 90+%
VONKRKBGTZDZNV-UHFFFAOYSA-N
ethylidenecyclopentane
75068
≥90%
2146-37-4
0.8
C7H12
MFCD00019319
UN3295
Soluble in water.
CC=C1CCCC1
96.173
96.17

Chemical Identifiers

2146-37-4
96.173
VONKRKBGTZDZNV-UHFFFAOYSA-N
75068
CC=C1CCCC1
C7H12
MFCD00019319
cyclopentane, ethylidene, ethyl idenecyclopentane, ethylidene-cyclopentane, cyclopentane,ethylidene, acmc-1cs2b, 1e-ethylidenecyclopentane, ethylidenecyclopentane, 90+%
ethylidenecyclopentane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224
Extremely flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 218-413-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only