CAS RN 2146-37-4
IUPAC Name:
ethylidenecyclopentane
Synonyms:
cyclopentane, ethylidene-
Molecular Weight (g/mol):
96.17
Molecular Formula:
C7H12
InChi Key:
VONKRKBGTZDZNV-UHFFFAOYSA-N
SMILES:
CC=C1CCCC1
1
–
1
of
1
results
1
Promotions Available
Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals
Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals
CAS: 2146-37-4 Molecular Formula: C7H12 Molecular Weight (g/mol): 96.173 MDL Number: MFCD00019319 InChI Key: VONKRKBGTZDZNV-UHFFFAOYSA-N Synonym: cyclopentane, ethylidene,ethyl idenecyclopentane,ethylidene-cyclopentane,cyclopentane,ethylidene,acmc-1cs2b,1e-ethylidenecyclopentane,ethylidenecyclopentane, 90+% PubChem CID: 75068 IUPAC Name: ethylidenecyclopentane SMILES: CC=C1CCCC1