missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Disodium Salt, Standard Solution (0.1M/Certified), Fisher Chemical

Catalog No. SS4121
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SS4121 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SS4121 Supplier Thermo Fisher Scientific Supplier No. FLSS4121
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6381-92-6 , 7732-18-5
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
Boiling Point 100°C
Melting Point 0°C
Concentration 0.0995 to 0.1005M
Density 1.02g/cm³
Grade Certified
Packaging Poly Bottle
Vapor Pressure 14mmHg
pH 4.4 to 4.5
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, Disodium Salt, Standard Solution, 0.1M
Show More Show Less