missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediamine-N,N'-diacetic Acid 98.0+%, TCI America™
Supplier: TCI America E027525G
Specifications
| Ethylenediamine-N,N′-diacetic Acid | |
| 200°C | |
| C6H10N2Na2O4 | |
| 25 g | |
| QMIFISRIAJBCGZ-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate | |
| 61975 | |
| ≥98.0% (T) |
| 5657-17-0 | |
| White | |
| MFCD00004281 | |
| edda, ethylenediamine-n,n'-diacetic acid, n,n'-ethylenediglycine, glycine, n,n'-1,2-ethanediylbis, ethylenediaminediacetic acid, glycine, n,n'-ethylenedi, unii-4lx0u5342u, 2-2-carboxymethylamino ethylamino acetic acid, ethylenediamine-n,n'-diaceticacid, ethylenediamine-n,n'-di acetic acid | |
| [Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O | |
| 220.14 | |
| 176.17 | |
| Crystalline Powder |
Chemical Identifiers
| 5657-17-0 | |
| 220.14 | |
| QMIFISRIAJBCGZ-UHFFFAOYSA-L | |
| 61975 | |
| [Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O |
| C6H10N2Na2O4 | |
| MFCD00004281 | |
| edda, ethylenediamine-n,n'-diacetic acid, n,n'-ethylenediglycine, glycine, n,n'-1,2-ethanediylbis, ethylenediaminediacetic acid, glycine, n,n'-ethylenedi, unii-4lx0u5342u, 2-2-carboxymethylamino ethylamino acetic acid, ethylenediamine-n,n'-diaceticacid, ethylenediamine-n,n'-di acetic acid | |
| disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate |
Safety and Handling
TSCA : Yes