missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine-N,N'-diacetic Acid 98.0+%, TCI America™

Catalog No. p-7128915
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02755G 5 g
E027525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E02755G Supplier TCI America Supplier No. E02755G
Only null left

Chemical Identifiers

CAS 5657-17-0
Molecular Formula C6H10N2Na2O4
Molecular Weight (g/mol) 220.14
MDL Number MFCD00004281
InChI Key QMIFISRIAJBCGZ-UHFFFAOYSA-L
Synonym edda, ethylenediamine-n,n'-diacetic acid, n,n'-ethylenediglycine, glycine, n,n'-1,2-ethanediylbis, ethylenediaminediacetic acid, glycine, n,n'-ethylenedi, unii-4lx0u5342u, 2-2-carboxymethylamino ethylamino acetic acid, ethylenediamine-n,n'-diaceticacid, ethylenediamine-n,n'-di acetic acid
PubChem CID 61975
IUPAC Name disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate
SMILES [Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O

Specifications

Melting Point 200°C
Color White
Quantity 5 g
Formula Weight 176.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediamine-N,N′-diacetic Acid