accessibility menu, dialog, popup

UserName

CAS RN 5657-17-0

NaNaOOHNNHOO

CAS RN 5657-17-0

IUPAC Name: disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate
Synonyms: disodium 2-({2-[(carboxylatomethyl)amino]ethyl}amino)acetate
Molecular Weight (g/mol): 220.14
Molecular Formula: C6H10N2Na2O4
InChi Key: QMIFISRIAJBCGZ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)