missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol/IPA/Water, 40/40/20, Reagents

Catalog No. p-200021792
Change view
Click to view available options
Quantity:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335367 10 L
01335366 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335367 Supplier Labchem Inc Supplier No. CE24700010F
Only null left

Ethylene Glycol/IPA/Water, 40/40/20, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 107-21-1 , 67-63-0
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym Antifreeze
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

Color Colorless
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 0.9613
Grade Reagent
Chemical Name or Material Ethylene Glycol/IPA/Water, 40/40/20