missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Diphenyl Ether 99.0+%, TCI America™

Catalog No. D08665G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D08665G Supplier TCI America Supplier No. D08665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104-66-5
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00003039
InChI Key XCSGHNKDXGYELG-UHFFFAOYSA-N
Synonym 1,2-diphenoxyethane, ethylene glycol diphenyl ether, diphenyl glycol, ethane, 1,2-diphenoxy, benzene, 1,1'-1,2-ethanediylbis oxy bis, 2-phenoxyethyl phenyl ether, unii-v9w31hx4aw, diphenyl cellosolve, 1,1'-ethane-1,2-diylbis oxy dibenzene, 2-phenoxyethoxy benzene
PubChem CID 7713
IUPAC Name (2-phenoxyethoxy)benzene
SMILES C(COC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 97°C
Color White-Yellow
Boiling Point 185°C
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Glycol Diphenyl Ether
EINECSNumber (3)-4146
TSCA Yes