CAS RN 104-66-5
IUPAC Name:
(2-phenoxyethoxy)benzene
Synonyms:
1,2-diphenoxyethane
Molecular Weight (g/mol):
214.26
Molecular Formula:
C14H14O2
InChi Key:
XCSGHNKDXGYELG-UHFFFAOYSA-N
SMILES:
C(COC1=CC=CC=C1)OC1=CC=CC=C1
1
–
1
of
1
results
1
Ethylene Glycol Diphenyl Ether 99.0+%, TCI America™
CAS: 104-66-5 Molecular Formula: C14H14O2 Molecular Weight (g/mol): 214.26 MDL Number: MFCD00003039 InChI Key: XCSGHNKDXGYELG-UHFFFAOYSA-N Synonym: 1,2-diphenoxyethane,ethylene glycol diphenyl ether,diphenyl glycol,ethane, 1,2-diphenoxy,benzene, 1,1'-1,2-ethanediylbis oxy bis,2-phenoxyethyl phenyl ether,unii-v9w31hx4aw,diphenyl cellosolve,1,1'-ethane-1,2-diylbis oxy dibenzene,2-phenoxyethoxy benzene PubChem CID: 7713 IUPAC Name: (2-phenoxyethoxy)benzene SMILES: C(COC1=CC=CC=C1)OC1=CC=CC=C1