missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Di(m-tolyl) Ether 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 98°C |
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 3077 |
| Formula Weight | 242.32 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethylene Glycol Di(m-tolyl) Ether |
Safety and Handling
| EINECSNumber | (3)-3886 |
| TSCA | Yes |