missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis[4-(ethoxycarbonyl)phenyl] Ether 98.0+%, TCI America™

Catalog No. E04145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04145G Supplier TCI America Supplier No. E04145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25909-66-4
Molecular Formula C20H22O6
Molecular Weight (g/mol) 358.39
MDL Number MFCD00144288
InChI Key XTHNYIOBLBKRMO-UHFFFAOYSA-N
Synonym 1,2-Bis(4-carbethoxyphenoxy)ethane, 1,2-Bis(4-ethoxycarbonylphenoxy)ethane, Ethylene Glycol Bis(4-carbethoxyphenyl) Ether
PubChem CID 11371768
IUPAC Name ethyl 4-{2-[4-(ethoxycarbonyl)phenoxy]ethoxy}benzoate
SMILES CCOC(=O)C1=CC=C(OCCOC2=CC=C(C=C2)C(=O)OCC)C=C1

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Formula Weight 358.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Glycol Bis[4-(ethoxycarbonyl)phenyl] Ether