CAS RN 25909-66-4
IUPAC Name:
ethyl 4-{2-[4-(ethoxycarbonyl)phenoxy]ethoxy}benzoate
Synonyms:
ethyl 4-{2-[4-(ethoxycarbonyl)phenoxy]ethoxy}benzoate
Molecular Weight (g/mol):
358.39
Molecular Formula:
C20H22O6
InChi Key:
XTHNYIOBLBKRMO-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=C(OCCOC2=CC=C(C=C2)C(=O)OCC)C=C1
1
–
1
of
1
results
1
Ethylene Glycol Bis[4-(ethoxycarbonyl)phenyl] Ether 98.0+%, TCI America™
CAS: 25909-66-4 Molecular Formula: C20H22O6 Molecular Weight (g/mol): 358.39 MDL Number: MFCD00144288 InChI Key: XTHNYIOBLBKRMO-UHFFFAOYSA-N Synonym: 1,2-Bis(4-carbethoxyphenoxy)ethane, 1,2-Bis(4-ethoxycarbonylphenoxy)ethane, Ethylene Glycol Bis(4-carbethoxyphenyl) Ether PubChem CID: 11371768 IUPAC Name: ethyl 4-{2-[4-(ethoxycarbonyl)phenoxy]ethoxy}benzoate SMILES: CCOC(=O)C1=CC=C(OCCOC2=CC=C(C=C2)C(=O)OCC)C=C1