Learn More
Ethylene glycol, 99.8%, anhydrous, AcroSeal™
CAS: 107-21-1 | C2H6O2 | 62.06 g/mol
Supplier: Thermo Scientific Chemicals 433810010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethylene glycol | |
| -13.0°C | |
| 196.0°C to 198.0°C | |
| Authentic | |
| AcroSeal™ Glass bottle | |
| 1.4300 to 1.4330 | |
| 1 L | |
| 15,3852 | |
| Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons | |
| C(CO)O | |
| 62.06 | |
| 21 mPa.s (20°C) | |
| 62.06 |
| 107-21-1 | |
| 1.1130g/mL | |
| 116°C | |
| 99.75% min. (GC) | |
| C2H6O2 | |
| HOCH2CH2OH | |
| 1.113 | |
| ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| 174 | |
| CHEBI:30742 | |
| 99.8% |
Chemical Identifiers
| 107-21-1 | |
| 62.06 | |
| ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
| CHEBI:30742 | |
| C(CO)O |
| C2H6O2 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| 174 | |
| ethane-1,2-diol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Warning
EINECSNumber : 203-473-3
RUO – Research Use Only