missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 99.8%, anhydrous, AcroSeal™

Catalog No. p-7077273
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC433811000 100 mL
AC433810010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433811000 Supplier Thermo Scientific Chemicals Supplier No. 433811000
Only null left

CAS: 107-21-1 | C2H6O2 | 62.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.06
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13.0°C
Density 1.1130g/mL
Boiling Point 196.0°C to 198.0°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4300 to 1.4330
Linear Formula HOCH2CH2OH
Quantity 100 mL
Merck Index 15,3852
Specific Gravity 1.113
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
Viscosity 21 mPa.s (20°C)
Formula Weight 62.06
Percent Purity 99.8%
Chemical Name or Material Ethylene glycol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 203-473-3

RUO – Research Use Only