missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, 40% v/v, Reagents

Catalog No. p-200021292
Change view
Click to view available options
Quantity:
10 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335369 10 L
01335368 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335369 Supplier Labchem Inc Supplier No. CE24600010F
Only null left

Ethylene Glycol, 40% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 107-21-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 1,2-Ethanediol, Monoethylene Glycol, Antifreeze
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 104°C
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0563
Percent Purity 40%
Grade Reagent
Chemical Name or Material Ethylene Glycol, 40% v/v