missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, 40% v/v, Reagents

Ethylene Glycol, 40% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CE246000200E

 View more versions of this product

Catalog No. 01-335-368

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethylene Glycol, 40% v/v
Colorless
200 L
Liquid
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
40%
7732-18-5,107-21-1
104°C
Poly Drum
1.0563
1,2-Ethanediol, Monoethylene Glycol, Antifreeze
O
18.02
Reagent

Chemical Identifiers

7732-18-5
18.02
1,2-Ethanediol, Monoethylene Glycol, Antifreeze
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions