missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol 1,2-Bis(2-propynyl) Ether 97.0+%, TCI America™
$397.05 - $1373.81
Chemical Identifiers
| CAS | 40842-04-4 |
|---|---|
| Molecular Formula | C8H10O2 |
| Molecular Weight (g/mol) | 138.166 |
| InChI Key | RNHWCSPBGKGOLM-UHFFFAOYSA-N |
| Synonym | 1,2-Bis(2-propynyloxy)ethane, 4,7-Dioxa-1,9-decadiyne |
| PubChem CID | 11083951 |
| IUPAC Name | 3-(2-prop-2-ynoxyethoxy)prop-1-yne |
| SMILES | C#CCOCCOCC#C |
Chemical Identifiers
| 40842-04-4 | |
| 138.166 | |
| 1,2-Bis(2-propynyloxy)ethane, 4,7-Dioxa-1,9-decadiyne | |
| 3-(2-prop-2-ynoxyethoxy)prop-1-yne |
| C8H10O2 | |
| RNHWCSPBGKGOLM-UHFFFAOYSA-N | |
| 11083951 | |
| C#CCOCCOCC#C |
Specifications
| 40842-04-4 | |
| 88°C | |
| 5 g | |
| 1,2-Bis(2-propynyloxy)ethane, 4,7-Dioxa-1,9-decadiyne | |
| C#CCOCCOCC#C | |
| 138.166 | |
| 138.17 | |
| Liquid |
| Yellow | |
| C8H10O2 | |
| 3271 | |
| RNHWCSPBGKGOLM-UHFFFAOYSA-N | |
| 3-(2-prop-2-ynoxyethoxy)prop-1-yne | |
| 11083951 | |
| ≥97.0% (GC) | |
| Ethylene Glycol 1,2-Bis(2-propynyl) Ether |
Safety and Handling
TSCA : No