missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol 1,2-Bis(2-propynyl) Ether 97.0+%, TCI America™
Supplier: TCI America E105425G
Specifications
| Ethylene Glycol 1,2-Bis(2-propynyl) Ether | |
| Yellow | |
| C8H10O2 | |
| 3271 | |
| RNHWCSPBGKGOLM-UHFFFAOYSA-N | |
| 3-(2-prop-2-ynoxyethoxy)prop-1-yne | |
| 11083951 | |
| ≥97.0% (GC) |
| 40842-04-4 | |
| 88°C | |
| 25 g | |
| 1,2-Bis(2-propynyloxy)ethane, 4,7-Dioxa-1,9-decadiyne | |
| C#CCOCCOCC#C | |
| 138.166 | |
| 138.17 | |
| Liquid |
Chemical Identifiers
| 40842-04-4 | |
| 138.166 | |
| 1,2-Bis(2-propynyloxy)ethane, 4,7-Dioxa-1,9-decadiyne | |
| 3-(2-prop-2-ynoxyethoxy)prop-1-yne |
| C8H10O2 | |
| RNHWCSPBGKGOLM-UHFFFAOYSA-N | |
| 11083951 | |
| C#CCOCCOCC#C |
Safety and Handling
TSCA : No