accessibility menu, dialog, popup

UserName

HCOOCH

CAS RN 40842-04-4

IUPAC Name: 3-[2-(prop-2-yn-1-yloxy)ethoxy]prop-1-yne
Synonyms: 3-[2-(prop-2-yn-1-yloxy)ethoxy]prop-1-yne
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: RNHWCSPBGKGOLM-UHFFFAOYSA-N
SMILES: C#CCOCCOCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results