Learn More
Ethylene chlorophosphite, 97%
CAS: 822-39-9 | C2H4ClO2P | 126.476 g/mol
Supplier: Thermo Scientific Chemicals A1272414
Description
Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.
Notes
Store in cool place. Moisture sensitive. Incompatible with water.
Specifications
| Ethylene chlorophosphite | |
| 1.415 | |
| 86°C (186°F) | |
| 1.491 | |
| 25 g | |
| 506179 | |
| ethylene chlorophosphite, 1,3,2-dioxaphospholane, 2-chloro, ethylene cyclo-chlorophosphite, acmc-209pnx, 1,3,2-dioxaphospholane,2-chloro, 2-chloro-1,3,2-dioxaphospholane, ethylene phosphorochloridite 2-chloro-1,3,2-dioxap, inchi=1/c2h4clo2p/c3-6-4-1-2-5-6/h1-2h | |
| C1COP(O1)Cl | |
| 126.476 | |
| 126.47 |
| 822-39-9 | |
| 46°C to 47°C (15 mmHg) | |
| C2H4ClO2P | |
| MFCD00014511 | |
| UN3265 | |
| Moisture sensitive | |
| OLSFRDLMFAOSIA-UHFFFAOYSA-N | |
| 2-chloro-1,3,2-dioxaphospholane | |
| 69973 | |
| 97% |
Chemical Identifiers
| 822-39-9 | |
| 126.476 | |
| OLSFRDLMFAOSIA-UHFFFAOYSA-N | |
| 2-chloro-1,3,2-dioxaphospholane |
| C2H4ClO2P | |
| MFCD00014511 | |
| 69973 | |
| C1COP(O1)Cl |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 212-499-4
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only