accessibility menu, dialog, popup

UserName

CAS RN 822-39-9

IUPAC Name: 2-chloro-1,3,2-dioxaphospholane
Synonyms: 2-chloro-1,3,2-dioxaphospholane
Molecular Weight (g/mol): 126.48
Molecular Formula: C2H4ClO2P
InChi Key: OLSFRDLMFAOSIA-UHFFFAOYSA-N
SMILES: ClP1OCCO1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)