missing translation for 'onlineSavingsMsg'
Learn More

Ethylene chlorophosphite, 97%

Catalog No. p-7020683
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1272414 25 g
AAA1272422 100 g
AAA1272436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1272414 Supplier Thermo Scientific Chemicals Supplier No. A1272414
Only null left

CAS: 822-39-9 | C2H4ClO2P | 126.476 g/mol

Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.

Notes
Store in cool place. Moisture sensitive. Incompatible with water.

Chemical Identifiers

CAS 822-39-9
Molecular Formula C2H4ClO2P
Molecular Weight (g/mol) 126.476
MDL Number MFCD00014511
InChI Key OLSFRDLMFAOSIA-UHFFFAOYSA-N
PubChem CID 69973
IUPAC Name 2-chloro-1,3,2-dioxaphospholane
SMILES C1COP(O1)Cl

Specifications

Density 1.415
Boiling Point 46°C to 47°C (15 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.491
Quantity 25 g
UN Number UN3265
Beilstein 506179
Sensitivity Moisture sensitive
Synonym ethylene chlorophosphite, 1,3,2-dioxaphospholane, 2-chloro, ethylene cyclo-chlorophosphite, acmc-209pnx, 1,3,2-dioxaphospholane,2-chloro, 2-chloro-1,3,2-dioxaphospholane, ethylene phosphorochloridite 2-chloro-1,3,2-dioxap, inchi=1/c2h4clo2p/c3-6-4-1-2-5-6/h1-2h
Formula Weight 126.47
Percent Purity 97%
Chemical Name or Material Ethylene chlorophosphite
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-499-4
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only