missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate 98.0+%, TCI America™

Supplier:  TCI America E099025G

 View more versions of this product

Catalog No. E099025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate
163°C
C8H11N3O3S
25 g
POBMBNPEUPDXRS-WDZFZDKYSA-N
ethyl (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetate
5582977
≥98.0% (HPLC,T)
64485-88-7
White-Yellow
MFCD00071532
ethyl 2z-2-amino-1,3-thiazol-4-yl methoxyimino ethanoate, ethyl-2-methoxy-amino-2-2-amino-4-thiazole-4-yl acetate, 2-amino-thiazol-4-yl-methoxyimino-acetic acid ethyl ester, z-ethyl 2-2-aminothiazol-4-yl-2-methoxyimino acetate, ethyl 2-amino-alpha-methoxyimino-4-thiazoleacetate, ethyl 2-amino-alpha-methoxyimino thiazol-4-acetate, ethyl 2z-2-2-amino-1,3-thiazol-4-yl-2-methoxyimino acetate, ethyl 2z-2-2-amino-1,3-thiazol-4-yl-2-methoxyiminoacetate, 2-amino-alpha-methoxyimino-4-thiazoleacetic acid ethyl ester, ethyl z-2-amino-alpha-methoxyimino thiazol-4-acetate
CCOC(=O)C(=NOC)C1=CSC(=N1)N
229.254
229.25
Crystalline Powder

Chemical Identifiers

64485-88-7
229.254
POBMBNPEUPDXRS-WDZFZDKYSA-N
5582977
CCOC(=O)C(=NOC)C1=CSC(=N1)N
C8H11N3O3S
MFCD00071532
ethyl 2z-2-amino-1,3-thiazol-4-yl methoxyimino ethanoate, ethyl-2-methoxy-amino-2-2-amino-4-thiazole-4-yl acetate, 2-amino-thiazol-4-yl-methoxyimino-acetic acid ethyl ester, z-ethyl 2-2-aminothiazol-4-yl-2-methoxyimino acetate, ethyl 2-amino-alpha-methoxyimino-4-thiazoleacetate, ethyl 2-amino-alpha-methoxyimino thiazol-4-acetate, ethyl 2z-2-2-amino-1,3-thiazol-4-yl-2-methoxyimino acetate, ethyl 2z-2-2-amino-1,3-thiazol-4-yl-2-methoxyiminoacetate, 2-amino-alpha-methoxyimino-4-thiazoleacetic acid ethyl ester, ethyl z-2-amino-alpha-methoxyimino thiazol-4-acetate
ethyl (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions