missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate 98.0+%, TCI America™

Catalog No. p-7129268
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09905G 5 g
E099025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09905G Supplier TCI America Supplier No. E09905G

Chemical Identifiers

CAS 64485-88-7
Molecular Formula C8H11N3O3S
Molecular Weight (g/mol) 229.254
MDL Number MFCD00071532
InChI Key POBMBNPEUPDXRS-WDZFZDKYSA-N
Synonym ethyl 2z-2-amino-1,3-thiazol-4-yl methoxyimino ethanoate, ethyl-2-methoxy-amino-2-2-amino-4-thiazole-4-yl acetate, 2-amino-thiazol-4-yl-methoxyimino-acetic acid ethyl ester, z-ethyl 2-2-aminothiazol-4-yl-2-methoxyimino acetate, ethyl 2-amino-alpha-methoxyimino-4-thiazoleacetate, ethyl 2-amino-alpha-methoxyimino thiazol-4-acetate, ethyl 2z-2-2-amino-1,3-thiazol-4-yl-2-methoxyimino acetate, ethyl 2z-2-2-amino-1,3-thiazol-4-yl-2-methoxyiminoacetate, 2-amino-alpha-methoxyimino-4-thiazoleacetic acid ethyl ester, ethyl z-2-amino-alpha-methoxyimino thiazol-4-acetate
PubChem CID 5582977
IUPAC Name ethyl (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetate
SMILES CCOC(=O)C(=NOC)C1=CSC(=N1)N

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 5 g
Formula Weight 229.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetate