missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Vanillate 98.0+%, TCI America™
$130.92 - $130.92
Chemical Identifiers
| CAS | 617-05-0 |
|---|---|
| Molecular Formula | C10H12O4 |
| Molecular Weight (g/mol) | 196.20 |
| MDL Number | MFCD00017269 |
| InChI Key | MWAYRGBWOVHDDZ-UHFFFAOYSA-N |
| Synonym | ethyl vanillate, vanillic acid, ethyl ester, 4-hydroxy-3-methoxybenzoic acid ethyl ester, benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester, unii-v38jk4z93o, 3-methoxy-4-hydroxybenzoic acid, ethyl ester, m-anisic acid, 4-hydroxy-, ethyl ester, vanilicacidethylester, ethyl 4-hydroxy-3-methoxy-benzoate, ethylvanillate |
| PubChem CID | 12038 |
| IUPAC Name | ethyl 4-hydroxy-3-methoxybenzoate |
| SMILES | CCOC(=O)C1=CC=C(O)C(OC)=C1 |
Chemical Identifiers
| 617-05-0 | |
| 196.20 | |
| MWAYRGBWOVHDDZ-UHFFFAOYSA-N | |
| 12038 | |
| CCOC(=O)C1=CC=C(O)C(OC)=C1 |
| C10H12O4 | |
| MFCD00017269 | |
| ethyl vanillate, vanillic acid, ethyl ester, 4-hydroxy-3-methoxybenzoic acid ethyl ester, benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester, unii-v38jk4z93o, 3-methoxy-4-hydroxybenzoic acid, ethyl ester, m-anisic acid, 4-hydroxy-, ethyl ester, vanilicacidethylester, ethyl 4-hydroxy-3-methoxy-benzoate, ethylvanillate | |
| ethyl 4-hydroxy-3-methoxybenzoate |
Specifications
| 617-05-0 | |
| White-Yellow | |
| C10H12O4 | |
| 25 g | |
| MWAYRGBWOVHDDZ-UHFFFAOYSA-N | |
| ethyl 4-hydroxy-3-methoxybenzoate | |
| 12038 | |
| ≥98.0% (GC) | |
| Ethyl Vanillate |
| 43°C | |
| 293°C | |
| MFCD00017269 | |
| ethyl vanillate, vanillic acid, ethyl ester, 4-hydroxy-3-methoxybenzoic acid ethyl ester, benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester, unii-v38jk4z93o, 3-methoxy-4-hydroxybenzoic acid, ethyl ester, m-anisic acid, 4-hydroxy-, ethyl ester, vanilicacidethylester, ethyl 4-hydroxy-3-methoxy-benzoate, ethylvanillate | |
| CCOC(=O)C1=CC=C(O)C(OC)=C1 | |
| 196.20 | |
| 196.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1560
RTECSNumber : YW5425000
TSCA : Yes