missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Vanillate 98.0+%, TCI America™

Catalog No. V005125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V005125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V005125G Supplier TCI America Supplier No. V005125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 617-05-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00017269
InChI Key MWAYRGBWOVHDDZ-UHFFFAOYSA-N
Synonym ethyl vanillate, vanillic acid, ethyl ester, 4-hydroxy-3-methoxybenzoic acid ethyl ester, benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester, unii-v38jk4z93o, 3-methoxy-4-hydroxybenzoic acid, ethyl ester, m-anisic acid, 4-hydroxy-, ethyl ester, vanilicacidethylester, ethyl 4-hydroxy-3-methoxy-benzoate, ethylvanillate
PubChem CID 12038
IUPAC Name ethyl 4-hydroxy-3-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C(OC)=C1

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 293°C
Quantity 25 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Vanillate
EINECSNumber (3)-1560
RTECSNumber YW5425000
TSCA Yes