missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vanillate, 97%

Catalog No. AAL0579809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0579809 10 g
AAL0579818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0579809 Supplier Thermo Scientific Chemicals Supplier No. L0579809

CAS: 617-05-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 617-05-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00017269
InChI Key MWAYRGBWOVHDDZ-UHFFFAOYSA-N
Synonym ethyl vanillate, vanillic acid, ethyl ester, 4-hydroxy-3-methoxybenzoic acid ethyl ester, benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester, unii-v38jk4z93o, 3-methoxy-4-hydroxybenzoic acid, ethyl ester, m-anisic acid, 4-hydroxy-, ethyl ester, vanilicacidethylester, ethyl 4-hydroxy-3-methoxy-benzoate, ethylvanillate
PubChem CID 12038
IUPAC Name ethyl 4-hydroxy-3-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C(OC)=C1

Specifications

Melting Point 42°C to 46°C
Boiling Point 293°C
Quantity 10 g
Beilstein 2100025
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material Ethyl vanillate
EINECSNumber 210-503-9
RTECSNumber YW5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only