missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (Triphenylphosphoranylidene)acetate 98.0+%, TCI America™

Supplier:  TCI America T1944250G

 View more versions of this product

Catalog No. T1944250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl (Triphenylphosphoranylidene)acetate
130°C
MFCD00009183
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
348.38
348.38
Crystalline Powder
1099-45-2
C22H21O2P
250 g
IIHPVYJPDKJYOU-UHFFFAOYSA-N
ethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
70670
≥98.0% (HPLC)

Chemical Identifiers

1099-45-2
348.38
IIHPVYJPDKJYOU-UHFFFAOYSA-N
70670
CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C22H21O2P
MFCD00009183
carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
ethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions