missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (Triphenylphosphoranylidene)acetate 98.0+%, TCI America™

Catalog No. p-7137033
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T194425G 25 g
T1944250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T194425G Supplier TCI America Supplier No. T194425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1099-45-2
Molecular Formula C22H21O2P
Molecular Weight (g/mol) 348.38
MDL Number MFCD00009183
InChI Key IIHPVYJPDKJYOU-UHFFFAOYSA-N
Synonym carbethoxymethylene triphenylphosphorane, ethyl triphenylphosphoranylidene acetate, triphenylcarbethoxymethylenephosphorane, acetic acid, triphenylphosphoranylidene-, ethyl ester, ethyl 2-triphenylphosphoranylidene acetate, ethoxycarbonylmethylene triphenylphosphorane, carboethoxymethylidenetriphenylphosphorane, ethyl triphenylphosphoranilidene acetate, carbethoxytriphenylphosphonium methylide, carbethoxymethylidenetriphenylphosphorane
PubChem CID 70670
IUPAC Name ethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
SMILES CCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 130°C
Quantity 25 g
Formula Weight 348.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl (Triphenylphosphoranylidene)acetate
TSCA Yes
Recommended Storage Refrigerator