missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Thiocyanate 98.0+%, TCI America™
Supplier: TCI America T0200500ML
Specifications
| Ethyl Thiocyanate | |
| Colorless | |
| C3H5NS | |
| 500 mL | |
| ethylthiocyanate, thiocyanic acid, ethyl ester, ethyl rhodanate, ethyl sulfocyanate, aethylrhodanid, ethane, thiocyanato, ethylthiokyanat, aethylrhodanid german, ethylthiokyanat czech, ethyl rhodanide | |
| CCSC#N | |
| 87.14 | |
| 87.14 | |
| Liquid |
| 542-90-5 | |
| 145°C | |
| MFCD00001834 | |
| 1992 | |
| WFCLYEAZTHWNEH-UHFFFAOYSA-N | |
| ethyl thiocyanate | |
| 10968 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 542-90-5 | |
| 87.14 | |
| WFCLYEAZTHWNEH-UHFFFAOYSA-N | |
| 10968 | |
| CCSC#N |
| C3H5NS | |
| MFCD00001834 | |
| ethylthiocyanate, thiocyanic acid, ethyl ester, ethyl rhodanate, ethyl sulfocyanate, aethylrhodanid, ethane, thiocyanato, ethylthiokyanat, aethylrhodanid german, ethylthiokyanat czech, ethyl rhodanide | |
| ethyl thiocyanate |
Safety and Handling
RTECSNumber : XK9900000
TSCA : Yes